siRNA Details
| Gene Symbol: Rhoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106461.1 | NP_001099931.1 | 4, 5 | 676 | ||
| XM_006233074.1 | XP_006233136.1 | 4, 5 | 650 | |||
| GenBank mRNA | BC166849.1 | - | 4, 5 | 692 | 9088252 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhoc| Gene Symbol: Rhoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106461.1 | NP_001099931.1 | 4, 5 | 676 | ||
| XM_006233074.1 | XP_006233136.1 | 4, 5 | 650 | |||
| GenBank mRNA | BC166849.1 | - | 4, 5 | 692 | 9088252 | |