siRNA Details
| Gene Symbol: Ccl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019233.1 | NP_062106.1 | 2 | 137 | ||
| XM_006245164.2 | XP_006245226.1 | 1, 2 | 203 | |||
| GenBank mRNA | AF053312.1 | - | 2 | 138 | ||
| U90447.1 | - | 2 | 137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl20| Gene Symbol: Ccl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019233.1 | NP_062106.1 | 2 | 137 | ||
| XM_006245164.2 | XP_006245226.1 | 1, 2 | 203 | |||
| GenBank mRNA | AF053312.1 | - | 2 | 138 | ||
| U90447.1 | - | 2 | 137 | |||