siRNA Details
| Gene Symbol: Slc44a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013914.1 | NP_001013936.1 | 10 | 1161 | ||
| GenBank mRNA | BC079142.1 | - | 10 | 1161 | 7128044 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc44a3| Gene Symbol: Slc44a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013914.1 | NP_001013936.1 | 10 | 1161 | ||
| GenBank mRNA | BC079142.1 | - | 10 | 1161 | 7128044 | |