siRNA Details
| Gene Symbol: Dnajb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013076.1 | NP_001013094.1 | 6 | 1317 | ||
| XM_006233488.1 | XP_006233550.1 | 5 | 1299 | |||
| XM_008761550.1 | XP_008759772.1 | 3 | 1684 | |||
| GenBank mRNA | BC083638.1 | - | 6 | 1317 | 7134832 | |

Dnajb4
Genomic Map