siRNA Details
| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024359.1 | NP_077335.1 | 2, 3 | 243 | ||
| GenBank mRNA | AF057308.1 | - | 2, 3 | 243 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hif1a| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024359.1 | NP_077335.1 | 2, 3 | 243 | ||
| GenBank mRNA | AF057308.1 | - | 2, 3 | 243 | ||