siRNA Details
| Gene Symbol: Arhgap15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013917.1 | NP_001013939.1 | 8, 9 | 767 | ||
| GenBank mRNA | BC079103.1 | - | 8, 9 | 767 | 7125019 | |
| FQ227834.1 | - | 5, 6 | 474 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap15| Gene Symbol: Arhgap15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013917.1 | NP_001013939.1 | 8, 9 | 767 | ||
| GenBank mRNA | BC079103.1 | - | 8, 9 | 767 | 7125019 | |
| FQ227834.1 | - | 5, 6 | 474 | |||