siRNA Details
| Gene Symbol: Crabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017244.1 | NP_058940.1 | 347 | |||
| GenBank mRNA | U23407.1 | - | 347 | |||
| Gene Symbol: LOC100911902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749317.3 | XP_003749365.1 | 2 | 1093 | ||

Crabp2
Genomic Map