siRNA Details
| Gene Symbol: Cybrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011954.1 | NP_001011954.1 | 2 | 374 | ||
| GenBank mRNA | BC085768.1 | - | 2 | 374 | 7108043 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cybrd1| Gene Symbol: Cybrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011954.1 | NP_001011954.1 | 2 | 374 | ||
| GenBank mRNA | BC085768.1 | - | 2 | 374 | 7108043 | |