siRNA Details
| Gene Symbol: Dnajc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106486.2 | NP_001099956.2 | 11 | 1300 | ||
| GenBank mRNA | BC100105.1 | - | 4 | 256 | ||
| FQ226920.1 | - | 10 | 911 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc10| Gene Symbol: Dnajc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106486.2 | NP_001099956.2 | 11 | 1300 | ||
| GenBank mRNA | BC100105.1 | - | 4 | 256 | ||
| FQ226920.1 | - | 10 | 911 | |||