siRNA Details
Gene Symbol: Frzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100527.1 | NP_001093997.1 | 4 | 951 | ||
XM_006234429.2 | XP_006234491.1 | 3 | 1375 | |||
GenBank mRNA | BC098777.1 | - | 1 | 49 | ||
BC166428.1 | - | 4 | 951 | 7106784 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
