siRNA Details
| Gene Symbol: Tcea2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057098.1 | NP_476439.1 | 1, 2 | 150 | ||
| XM_006235742.2 | XP_006235804.1 | 1, 2 | 201 | |||
| GenBank mRNA | D12927.1 | - | 1, 2 | 150 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcea2| Gene Symbol: Tcea2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057098.1 | NP_476439.1 | 1, 2 | 150 | ||
| XM_006235742.2 | XP_006235804.1 | 1, 2 | 201 | |||
| GenBank mRNA | D12927.1 | - | 1, 2 | 150 | ||