siRNA Details
| Gene Symbol: Pecam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031591.1 | NP_113779.1 | 1638 | |||
| GenBank mRNA | BC104711.1 | - | 1638 | 7368518 | ||
| U77697.1 | - | 597 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pecam1| Gene Symbol: Pecam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031591.1 | NP_113779.1 | 1638 | |||
| GenBank mRNA | BC104711.1 | - | 1638 | 7368518 | ||
| U77697.1 | - | 597 | ||||