siRNA Details
| Gene Symbol: Gjb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019241.1 | NP_062114.1 | 1 | 312 | ||
| XM_008763990.1 | XP_008762212.1 | 2 | 556 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gjb5| Gene Symbol: Gjb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019241.1 | NP_062114.1 | 1 | 312 | ||
| XM_008763990.1 | XP_008762212.1 | 2 | 556 | |||