siRNA Details
| Gene Symbol: Bcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017253.2 | NP_058949.1 | 2 | 154 | ||
| XM_008763370.1 | XP_008761592.1 | 2 | 167 | |||
| XM_008763371.1 | XP_008761593.1 | 2 | 417 | |||
| XM_008763372.1 | XP_008761594.1 | 3 | 243 | |||
| XM_008763373.1 | XP_008761595.1 | 2 | 346 | |||
| GenBank mRNA | AJ278701.1 | - | 2 | 148 | ||
| BC087710.1 | - | 2 | 154 | 7315507 | ||
| U35774.1 | - | 2 | 120 | |||

Bcat1
Genomic Map