siRNA Details
| Gene Symbol: Mettl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024981.1 | NP_001020152.1 | 5 | 876 | ||
| GenBank mRNA | BC091215.1 | - | 5 | 876 | 7383613 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl15| Gene Symbol: Mettl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024981.1 | NP_001020152.1 | 5 | 876 | ||
| GenBank mRNA | BC091215.1 | - | 5 | 876 | 7383613 | |