siRNA Details
| Gene Symbol: Galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013919.1 | NP_001013941.1 | 6, 7 | 675 | ||
| XM_006234904.2 | XP_006234966.1 | 5, 6 | 1130 | |||
| XM_006234906.2 | XP_006234968.1 | 5, 6 | 635 | |||
| GenBank mRNA | BC083716.1 | - | 6, 7 | 675 | 7190694 | |

Galk2
Genomic Map