siRNA Details
| Gene Symbol: Fam227b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013920.2 | NP_001013942.2 | 11 | 958 | ||
| XM_006234909.2 | XP_006234971.1 | 11 | 881 | |||
| XM_008762153.1 | XP_008760375.1 | 11 | 915 | |||
| GenBank mRNA | BC079426.1 | - | 11 | 917 | 7114265 | |
| BC089861.1 | - | 11 | 914 | 7114720 | ||

Fam227b
Genomic Map