siRNA Details
| Gene Symbol: Usp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106502.1 | NP_001099972.1 | 19 | 3386 | ||
| XM_006234916.2 | XP_006234978.1 | 19 | 3459 | |||
| XM_008762155.1 | XP_008760377.1 | 19 | 3459 | |||
| GenBank mRNA | FQ235333.1 | - | 5 | 766 | ||

Usp8
Genomic Map