siRNA Details
| Gene Symbol: Chchd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106509.1 | NP_001099979.1 | 2 | 182 | ||
| XM_006234969.2 | XP_006235031.1 | 2 | 227 | |||
| GenBank mRNA | BC158612.1 | - | 2 | 84 | 7384755 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chchd5| Gene Symbol: Chchd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106509.1 | NP_001099979.1 | 2 | 182 | ||
| XM_006234969.2 | XP_006235031.1 | 2 | 227 | |||
| GenBank mRNA | BC158612.1 | - | 2 | 84 | 7384755 | |