siRNA Details
Gene Symbol: Btg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017259.1 | NP_058955.1 | 1 | 157 | ||
GenBank mRNA | BC072493.1 | - | 1 | 116 | 7097327 | |
M60921.1 | - | 1 | 157 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Btg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017259.1 | NP_058955.1 | 1 | 157 | ||
GenBank mRNA | BC072493.1 | - | 1 | 116 | 7097327 | |
M60921.1 | - | 1 | 157 |