siRNA Details
| Gene Symbol: Ovol2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106519.1 | NP_001099989.1 | 4 | 536 | ||
| XM_006235124.2 | XP_006235186.1 | 4 | 1695 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ovol2| Gene Symbol: Ovol2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106519.1 | NP_001099989.1 | 4 | 536 | ||
| XM_006235124.2 | XP_006235186.1 | 4 | 1695 | |||