siRNA Details
Gene Symbol: Cstl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106522.1 | NP_001099992.1 | 3 | 421 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cstl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106522.1 | NP_001099992.1 | 3 | 421 |