siRNA Details
| Gene Symbol: Cst7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106523.1 | NP_001099993.1 | 3, 4 | 495 | ||
| XM_006235213.2 | XP_006235275.1 | 4, 5 | 1768 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cst7| Gene Symbol: Cst7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106523.1 | NP_001099993.1 | 3, 4 | 495 | ||
| XM_006235213.2 | XP_006235275.1 | 4, 5 | 1768 | |||