siRNA Details
Gene Symbol: Srxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047858.3 | NP_001041323.3 | 2 | 511 | ||
XM_008762308.1 | XP_008760530.1 | 3 | 306 | |||
GenBank mRNA | AY325205.1 | - | 3 | 306 | ||
BC167743.1 | - | 2 | 264 |
