siRNA Details
| Gene Symbol: Srxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047858.3 | NP_001041323.3 | 2 | 627 | ||
| GenBank mRNA | BC167743.1 | - | 2 | 380 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srxn1| Gene Symbol: Srxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047858.3 | NP_001041323.3 | 2 | 627 | ||
| GenBank mRNA | BC167743.1 | - | 2 | 380 | ||