siRNA Details
| Gene Symbol: Plagl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106528.1 | NP_001099998.1 | 3 | 594 | ||
| XM_006235285.2 | XP_006235347.1 | 3 | 638 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plagl2| Gene Symbol: Plagl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106528.1 | NP_001099998.1 | 3 | 594 | ||
| XM_006235285.2 | XP_006235347.1 | 3 | 638 | |||