siRNA Details
| Gene Symbol: Kif3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106529.1 | NP_001099999.1 | 2 | 901 | ||
| XM_006235287.2 | XP_006235349.1 | 2 | 1322 | |||
| XM_006235288.1 | XP_006235350.1 | 2 | 938 | |||
| XM_006235289.2 | XP_006235351.1 | 2 | 1118 | |||

Kif3b
Genomic Map