siRNA Details
Gene Symbol: Eif2s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199380.1 | NP_955412.1 | 9 | 946 | ||
GenBank mRNA | BC062402.1 | - | 9 | 946 | 5621303 | |
FQ209519.1 | - | 7 | 623 | |||
FQ213305.1 | - | 9 | 978 | |||
FQ215337.1 | - | 9 | 991 | |||
FQ215587.1 | - | 9 | 887 | |||
FQ234002.1 | - | 9 | 975 |
