siRNA Details
| Gene Symbol: Myl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100885.1 | NP_001094355.1 | 1 | 75 | ||
| GenBank mRNA | S77900.1 | - | 1 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myl9| Gene Symbol: Myl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100885.1 | NP_001094355.1 | 1 | 75 | ||
| GenBank mRNA | S77900.1 | - | 1 | 75 | ||