siRNA Details
| Gene Symbol: Ndrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013923.1 | NP_001013945.1 | 3, 4 | 254 | ||
| GenBank mRNA | BC078946.1 | - | 3, 4 | 254 | 7109582 | |
| Gene Symbol: LOC679863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762423.1 | XP_008760645.1 | 3, 4 | 299 | ||

Ndrg3
Genomic Map