siRNA Details
| Gene Symbol: Znfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047860.1 | NP_001041325.1 | 7 | 2315 | ||
| XM_006235641.1 | XP_006235703.1 | 4 | 2413 | |||
| XM_006235642.2 | XP_006235704.1 | 4 | 2153 | |||
| XM_006235643.2 | XP_006235705.1 | 4 | 2061 | |||
| GenBank mRNA | AY325141.1 | - | 7 | 2315 | ||

Znfx1
Genomic Map