siRNA Details
| Gene Symbol: Kcng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106545.1 | NP_001100015.1 | 2 | 525 | ||
| XM_008762484.1 | XP_008760706.1 | 2 | 344 | |||
| GenBank mRNA | M81784.1 | - | 2 | 350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcng1| Gene Symbol: Kcng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106545.1 | NP_001100015.1 | 2 | 525 | ||
| XM_008762484.1 | XP_008760706.1 | 2 | 344 | |||
| GenBank mRNA | M81784.1 | - | 2 | 350 | ||