siRNA Details
| Gene Symbol: Lkaaear1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106551.1 | NP_001100021.1 | 3, 4 | 557 | ||
| XM_006235763.2 | XP_006235825.1 | 3, 4 | 667 | |||
| XM_006235764.2 | XP_006235826.1 | 3, 4 | 606 | |||
| XM_008762501.1 | XP_008760723.1 | 3, 4 | 535 | |||
| XM_008762502.1 | XP_008760724.1 | 2, 3 | 529 | |||

Lkaaear1
Genomic Map