siRNA Details
| Gene Symbol: Lkaaear1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106551.1 | NP_001100021.1 | 2 | 104 | ||
| XM_006235763.2 | XP_006235825.1 | 2 | 214 | |||
| XM_006235764.2 | XP_006235826.1 | 2 | 153 | |||
| XM_008762501.1 | XP_008760723.1 | 2 | 82 | |||
| XM_008762502.1 | XP_008760724.1 | 1 | 76 | |||

Lkaaear1
Genomic Map