siRNA Details
| Gene Symbol: Dlx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100531.1 | NP_001094001.1 | 3 | 1098 | ||
| XM_008761894.1 | XP_008760116.1 | 2 | 809 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlx1| Gene Symbol: Dlx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100531.1 | NP_001094001.1 | 3 | 1098 | ||
| XM_008761894.1 | XP_008760116.1 | 2 | 809 | |||