siRNA Details
| Gene Symbol: Adam10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019254.1 | NP_062127.1 | 6 | 908 | ||
| XM_006243369.2 | XP_006243431.1 | 4 | 530 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam10| Gene Symbol: Adam10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019254.1 | NP_062127.1 | 6 | 908 | ||
| XM_006243369.2 | XP_006243431.1 | 4 | 530 | |||