siRNA Details
| Gene Symbol: Tubb4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199094.2 | NP_954525.1 | 4 | 507 | ||
| XM_006233582.2 | XP_006233644.1 | 4 | 520 | |||
| GenBank mRNA | BC060597.1 | - | 4 | 511 | 6889737 | |
| FQ213825.1 | - | 4 | 504 | |||
| FQ215022.1 | - | 4 | 504 | |||
| FQ227178.1 | - | 4 | 504 | |||
| FQ228801.1 | - | 4 | 503 | |||

Tubb4b
Genomic Map