siRNA Details
| Gene Symbol: Fut7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199491.1 | NP_955785.1 | 3 | 571 | ||
| XM_006233585.2 | XP_006233647.1 | 2 | 457 | |||
| XM_006233586.2 | XP_006233648.1 | 2 | 151 | |||
| XM_006233587.2 | XP_006233649.1 | 2 | 180 | |||
| XM_008761582.1 | XP_008759804.1 | 2 | 255 | |||
| XM_008761583.1 | XP_008759805.1 | 2 | 406 | |||
| GenBank mRNA | AJ276068.2 | - | 3 | 671 | ||

Fut7
Genomic Map