siRNA Details
| Gene Symbol: Tmem8c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134517.1 | NP_001127989.1 | 3 | 574 | ||
| XM_008761635.1 | XP_008759857.1 | 3 | 576 | |||
| XM_008761636.1 | XP_008759858.1 | 3 | 325 | |||
| XM_008761637.1 | XP_008759859.1 | 1 | 46 | |||

Tmem8c
Genomic Map