siRNA Details
| Gene Symbol: Psmb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057099.3 | NP_476440.2 | 5, 6 | 588 | ||
| GenBank mRNA | BC058451.1 | - | 5, 6 | 592 | 6891542 | |
| D10754.1 | - | 5, 6 | 577 | |||
| FQ209811.1 | - | 5, 6 | 591 | |||
| FQ217385.1 | - | 5, 6 | 591 | |||
| FQ224639.1 | - | 5, 6 | 591 | |||
| Gene Symbol: LOC100360846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729028.3 | XP_002729074.2 | 2 | 736 | ||

Psmb6
Genomic Map