siRNA Details
| Gene Symbol: Srsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195505.1 | NP_001182434.1 | 327 | |||
| NM_001195506.1 | NP_001182435.1 | 198 | ||||
| NM_019257.3 | NP_062130.2 | 202 | ||||
| XM_008764878.1 | XP_008763100.1 | 2 | 378 | |||
| XM_008764879.1 | XP_008763101.1 | 2 | 249 | |||
| GenBank mRNA | AF020683.1 | - | 2 | 264 | ||
| BC058479.1 | - | 196 | 6920786 | |||
| FM049719.1 | - | 2 | 290 | |||
| FM059586.1 | - | 2 | 176 | |||
| FM081441.1 | - | 2 | 202 | |||
| L13635.1 | - | 298 | ||||

Srsf5
Genomic Map