siRNA Details
| Gene Symbol: Srsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195505.1 | NP_001182434.1 | 752 | |||
| NM_001195506.1 | NP_001182435.1 | 623 | ||||
| NM_019257.3 | NP_062130.2 | 627 | ||||
| XM_008764878.1 | XP_008763100.1 | 7 | 803 | |||
| XM_008764879.1 | XP_008763101.1 | 7 | 674 | |||
| GenBank mRNA | AF020683.1 | - | 6 | 1924 | ||
| BC058479.1 | - | 621 | 6920786 | |||
| L13635.1 | - | 723 | ||||

Srsf5
Genomic Map