siRNA Details
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017279.1 | NP_058975.1 | 3, 4 | 253 | ||
| GenBank mRNA | FQ214776.1 | - | 3, 4 | 268 | ||
| FQ223872.1 | - | 268 | ||||
| J02897.1 | - | 3, 4 | 253 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psma2| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017279.1 | NP_058975.1 | 3, 4 | 253 | ||
| GenBank mRNA | FQ214776.1 | - | 3, 4 | 268 | ||
| FQ223872.1 | - | 268 | ||||
| J02897.1 | - | 3, 4 | 253 | |||