siRNA Details
| Gene Symbol: Crygn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106573.1 | NP_001100043.1 | 1, 2 | 90 | ||
| XM_006235882.2 | XP_006235944.1 | 2, 3 | 761 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crygn| Gene Symbol: Crygn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106573.1 | NP_001100043.1 | 1, 2 | 90 | ||
| XM_006235882.2 | XP_006235944.1 | 2, 3 | 761 | |||