siRNA Details
| Gene Symbol: Psma6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017283.3 | NP_058979.1 | 4 | 380 | ||
| XM_006240098.2 | XP_006240160.1 | 4 | 338 | |||
| GenBank mRNA | BC062232.1 | - | 4 | 381 | 6888689 | |
| D10755.1 | - | 4 | 369 | |||
| FQ212808.1 | - | 482 | ||||
| FQ220193.1 | - | 4 | 407 | |||

Psma6
Genomic Map