siRNA Details
| Gene Symbol: Armc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106576.2 | NP_001100046.2 | 4 | 704 | ||
| GenBank mRNA | BC079464.1 | - | 4 | 608 | ||
| BC161913.1 | - | 4 | 709 | 8369555 | ||
| DY320753.1 | - | 4 | 704 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc10| Gene Symbol: Armc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106576.2 | NP_001100046.2 | 4 | 704 | ||
| GenBank mRNA | BC079464.1 | - | 4 | 608 | ||
| BC161913.1 | - | 4 | 709 | 8369555 | ||
| DY320753.1 | - | 4 | 704 | |||