siRNA Details
| Gene Symbol: Cst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019258.1 | NP_062131.1 | 4 | 584 | ||
| GenBank mRNA | AF090692.1 | - | 4 | 584 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cst8| Gene Symbol: Cst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019258.1 | NP_062131.1 | 4 | 584 | ||
| GenBank mRNA | AF090692.1 | - | 4 | 584 | ||