siRNA Details
| Gene Symbol: Lsm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106585.1 | NP_001100055.1 | 1 | 157 | ||
| GenBank mRNA | BC167094.1 | - | 1 | 28 | 7462085 | |
| FQ211862.1 | - | 123 | ||||
| FQ212479.1 | - | 123 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsm8| Gene Symbol: Lsm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106585.1 | NP_001100055.1 | 1 | 157 | ||
| GenBank mRNA | BC167094.1 | - | 1 | 28 | 7462085 | |
| FQ211862.1 | - | 123 | ||||
| FQ212479.1 | - | 123 | ||||