siRNA Details
| Gene Symbol: Lsm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106585.1 | NP_001100055.1 | 3, 4 | 339 | ||
| GenBank mRNA | BC167094.1 | - | 3, 4 | 210 | 7462085 | |
| FQ211862.1 | - | 305 | ||||
| FQ212479.1 | - | 305 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsm8| Gene Symbol: Lsm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106585.1 | NP_001100055.1 | 3, 4 | 339 | ||
| GenBank mRNA | BC167094.1 | - | 3, 4 | 210 | 7462085 | |
| FQ211862.1 | - | 305 | ||||
| FQ212479.1 | - | 305 | ||||