siRNA Details
| Gene Symbol: Pla2g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M25148.1 | - | 4 | 311 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g2a| Gene Symbol: Pla2g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M25148.1 | - | 4 | 311 | ||